VWR® Cleanroom Mask with Ties, Made in USA
Supplier: VWR International
76409-318
76409-318CS 213 USD InStock
76409-318
VWR® Cleanroom Mask with Ties, Made in USA
Masks Cleanroom Masks
The combination of strong BFE and PFE make these masks ideal for use in production or research areas.
  • Protection against bacteria and particles
  • 3-ply construction
  • Non sterile
  • Tie configuration
A key part of the VWR® Made in USA PPE line, these cleanroom masks with ties provide a comfortable fit with the adjustability user's desire.
Country of origin: USA.
Información para pedidos: Made in USA Masks with earloops are also available; see catalog number 76451-956.
Envase: Masks are packaged 50 per double bag.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty