QuikVap™ Evaporating System, Environmental Express®
Supplier: Environmental Express
G7000
76436-990EA 4690.82 USD
76436-990
QuikVap™ Evaporating System, Environmental Express®
Evaporators Microevaporators
One system handles both flask and pan analysis methods.
  • Accepts both borosilicate glass flasks or aluminum evaporating pans--simply switch out adapter plate
  • Evaporate n-hexane quickly and effectively for oil and grease testing
  • Caution–operation should take place under an operating fume hood
Versatile system evaporates n-hexane quickly and effectively with adjustable temperature control, airflow control, and vessel configuration. Use digital controller to accurately set and maintain temperature throughout the evaporation process. System accepts either 1More Product Information