RNAdvance Viral RNA Extraction Kit, Beckman Coulter
Supplier: BURAM
C63510
76446-908EA 3327.77 USD InStock
76446-908
RNAdvance Viral RNA Extraction Kit, Beckman Coulter
Buffers Extraction Buffers
Extract high-quality RNA from saliva, nasopharyngeal and oropharyngeal swab collection devices.
  • Produces viral RNA of high integrity and purity for use in PCR-based assays and NGS
  • Performance Limit of Detection (LoD) demonstrated at 1 copy/µL for viral RNA
  • Compatible with multiple sample inputs, including saliva and swab transport media
  • Automation-ready chemistry adaptable to high-throughput workflows
  • Uses saliva and swab in universal transport media as the starting sample material
Powered by Solid Phase Reversible Immobilization (SPRI) technology, the RNAdvance Viral kit uses paramagnetic beads to selectively bind nucleic acids for high yield and efficient removal of impurities. The scalable protocol can be performed in single tubes or 96-well plates with the flexibility to automate on a variety of liquid handling platforms.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty