StrandBrite™ Green Fluorimetric RNA Quantitation Kit *High Selectivity*, AAT Bioquest
Supplier: AAT BIOQUEST INC
17657
76482-952EA 437.45 USD InStock
76482-952
StrandBrite™ Green Fluorimetric RNA Quantitation Kit *High Selectivity*, AAT Bioquest
Assays Nucleic Acid Assays
The major challenge to analyze RNA in live cells is the interferences caused by DNA.
  • Highly sensitive RNA detection
  • Robust protocol
  • Multiple applications
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty