PE-Cy7 Tandem, ReadiUse™ fluorescent probe, preactivated

Supplier: AAT BIOQUEST INC
Warning

2582
76484-144EA 437.45 USD
76484-144
PE-Cy7 Tandem, ReadiUse™ fluorescent probe, preactivated
PE-Cy7 Tandem

PE-Cy7 is a popular color used in flow cytometry.


  • Ready to use
  • Excellent conjugation yield

MW: 240000 g/mol
ADR: 5 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de03c959172e786803a2394c3475b4c9ece1c56f&vid=de03c959172e786803a2394c3475b4c9ece1c56f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR