ReadiLink™ Rapid XFD488 Antibody Labeling Kit *XFD488 Same Structure to Alexa Fluor™ 488* (2×50 µg Labelings), AAT Bioquest

Supplier: AAT BIOQUEST INC

1275
76485-560EA 177.52 USD
76485-560
ReadiLink™ Rapid XFD488 Antibody Labeling Kit *XFD488 Same Structure to Alexa Fluor™ 488* (2×50 µg Labelings), AAT Bioquest
Protein Linking and Labeling Systems

XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).


  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required


Precaución: For research use only.

Order Now

SPECIFICATIONS

Environmentally Preferable
Host

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR