Stainless Steel Cleanroom Waste Basket, Transforming Technologies
Supplier: Transforming Technologies
WBAS28MET
76490-262EA 74.68 USD
76490-262
Stainless Steel Cleanroom Waste Basket, Transforming Technologies
Bins Waste Bins
Conductive trash cans keep charged waste materials away from sensitive electronic devices and from chemicals that could be ignited by a spark.
  • Ideal for cleanrooms and other controlled environments
  • 7 gallon (28 quart) capacity
  • Made with permanently conductive stainless steel
  • Typical point-to-point resistance: 10² to 10³ ohm
When grounded, the cans and liners will not hold a static charge and will contain materials safely. This conductive ESD waste basket is a stylish rectangular wastebasket with radius corners and rolled edges. Stainless steelMore Product Information