Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mopec Bone Dust Vacuum for Swordfish Autopsy Saw, Mortech®

Supplier: Mortech Manufacturing
BD093 BD095
76498-350EA 7608.6 USD InStock
76498-350 76498-354
Mopec Bone Dust Vacuum for Swordfish Autopsy Saw, Mortech®
Vacuum Cleaners
This vacuum will extract harmful debris and matter from the air. Flexible, clear plastic hood improves visibility. Compatible with the Mopec Swordfish Autopsy Saw only.

  • Hand piece storage unit, improved stability
  • Three-stage filtration: HEPA filter, cloth filter, and paper filter bag
  • Easy access to filters
  • IP 44 water resistance
  • Bag capacity: 7 L (1.84 US gallon)
  • Top-mounted lead for easy removal
  • Air volume: 90 L/s

Compact and portable, the vacuum features four large casters all with brakes, and a stable base. This vacuum is specifically designed forMore Product Information
Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty