Scintillation Vials, Clear Borosilicate Glass, without Closures, Qorpak®
Supplier: Qorpak
256283
76540-004CS 272.8 USD InStock
76540-004
Scintillation Vials, Clear Borosilicate Glass, without Closures, Qorpak®
Vials
These scintillation vials fit all standard equipment for liquid scintillation systems. Manufactured with critical tolerances, ensuring reliable, consistent results in scintillation counting.
  • Clear Type I Borosilicate, 33 expansion glass construction
  • Fits all standard liquid scintillation counting equipment
  • High quality vials produced in accordance to industry standards
22 mm screw caps purchased separately, see Qorpak part# CAP-00156 or CAP-00162 for the most popular caps for this vial.
Envase: 4 packs of 144 per case of 576 vials.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty