Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Quinolinecarbaldehyde 98%

Supplier: AMBEED, INC
Warning

A147064-1G A147064-5G A147064-100G A147064-1G A147064-5G
103803-730EA 13 USD InStock
103803-730 103841-826 76798-466 77317-182 77317-184
8-Quinolinecarbaldehyde 98%
8-Quinolinecarbaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 95…96 °C
Flash Pt: 151.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00805837
CAS Number: 38707-70-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty