6-Bromopiperonal 98%

Supplier: AMBEED, INC
Warning

A735142-25G A735142-5G A735142-100G A735142-10G A735142-500G A735142-1G
103840-810EA 14 USD
103840-810 76845-036 76845-038 77105-128 77105-130 77105-132 77308-234
6-Bromopiperonal 98%
6-Bromopiperonal

Precaución: for Research Use Only

Formula: C₈H₅BrO₃
MW: 229.03 g/mol
Boiling Pt: 313.9 °C (760 mm Hg)
Melting Pt: 128…132 °C
Flash Pt: 143.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022952
CAS Number: 15930-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR