10X Citrate Buffer, pH 6.0
Supplier: AAT BIOQUEST INC
10000
77461-344EA 101.72 USD
77461-344
10X Citrate Buffer, pH 6.0
Buffers Biological Buffers
A citrate buffer is an aqueous solution composed of a weak acid (citric acid) and its conjugate base (sodium citrate).
  • Designed for ease of use
  • Saves time
Buffer solution is intended for heat-induced antigen retrieval in IHC.
Precaución: For research use only.