Recombinase Polymerase Amplification (RPA) Kit
Supplier: G-BIOSCIENCES
786-2155
77540-368EA 316.45 USD
77540-368
Recombinase Polymerase Amplification (RPA) Kit
Nucleic Acid Reagents Isothermal Amplification Reagents
A mixture of RPA enzymes (UvsX, UvsY, GP32 and Bsu) in liquid form.
  • Use for isothermal amplification
Rapid diagnostic assay.