Scorpion Scoop Eco Powder Scoop
Supplier: SAMPLING SYSTEMS LTD
A334-1000
77577-868EA 56.09 USD
77577-868
Scorpion Scoop Eco Powder Scoop
Scoops
The Scorpion Scoop Eco is a stainless steel powder scoop that is ideal for general industrial use.
  • Light duty design
  • Spot welded with a satin finish
  • Perfect for non hygienic applications
The Scorpion Scoop Eco is a light duty version of our Scorpion PharmaScoop. The handle on top makes scooping powder easy, similar to a traditional flour scoop.
The Scorpion Scoop Eco has a spot welded construction, this means that it is not suitable for hygienic applications in the pharmaceutical and food industries. For hygienic applications please choose a scoopMore Product Information