VWR® Advanced Research Microscopes
Supplier: VWR International
77676-446 77676-448
77676-446EA 3745 USD InStock
77676-446 77676-448
VWR® Advanced Research Microscopes
Microscopes
This microscope for diverse questions from life sciences, but also from material sciences.
  • Advanced compound microscope binocular head Siedentopf type 30°. inclined, 360° rotating
  • Widefield high eyepoint eyepieces WFPL10×/20 mm with diopter adjustment on both eyepieces, with rubber eyecup (paired)
  • Reversed sextuple nosepiece
  • CCIS Plan achromat objectives EC-H PL4×, EC-H PL10×, EC-H PL40× S, EC-H PL100× S-oil
  • Coaxial coarse and fine focusing adjustment
  • Built in low position rackless stage with right hand control
  • Achromat swing-out condenser N.A. 0.90 with
More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty