VWR® Microscopes with Camera Bundle, Professional Plus Trinocular Microscope with HDMI or Tablet Camera
Supplier: VWR International
77676-456 77676-458 77676-460
77676-456EA 4052 USD InStock
77676-456 77676-458 77676-460
VWR® Microscopes with Camera Bundle, Professional Plus Trinocular Microscope with HDMI or Tablet Camera
Microscopes
The microscope is for advanced work in life sciences. Sections, smears and dispersions can be checked by plan achromat objectives. Lab technicians profit from the Full-Kohler illumination, ready to work on unstained or weakly-stained structures from biology and medicine.
  • Compound microscope with trinocular head Siedentopf type 30° inclined, 360° swiveling, 360° rotating (light split 100:0/20:80)
  • Widefield high eyepoint eyepiece N-WF10×/20 mm with diopter adjustment on both eyepieces and rubber eyecups
  • Reversed quintuple revolving nosepiece
  • CCIS EC Plan
More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty