NanoXact Gold Nanospheres, 0.05 mg/ml (Au), Citrate Surface, nanoComposix
Supplier: NANOCOMPOSIX, INC.
AUCN20-500M AUCN30-100M AUCN30-500M AUCN40-100M AUCN40-500M AUCN50-100M AUCN50-500M AUCN60-100M AUCN100-100M AUCN100-500M AUCN5-500M AUCN10-100M AUCN10-500M AUCN20-100M AUCN60-500M AUCN80-100M AUCN80-500M AUCN5-100M AUCN100-500M AUCN100-50M AUCN10-100M AUCN10-500M AUCN10-50M AUCN20-100M AUCN20-500M AUCN20-50M AUCN30-100M AUCN30-500M AUCN30-50M AUCN40-100M AUCN40-500M AUCN40-50M AUCN50-100M AUCN80-100M AUCN80-500M AUCN80-50M AUCN100-100M AUCN50-500M AUCN50-50M AUCN5-100M AUCN5-500M AUCN5-50M AUCN60-100M AUCN60-500M AUCN60-50M
103369-358EA 755 USD
103369-358 103369-362 103369-364 103369-368 103369-370 103369-374 103369-376 103369-380 103369-384 103369-386 103369-806 103369-816 103369-818 103369-922 103369-924 103369-928 103369-930 103370-208 77679-962 77679-964 77679-966 77679-970 77679-972 77679-974 77679-978 77679-980 77679-982 77679-986 77679-988 77679-990 77679-994 77679-996 77679-998 77680-022 77680-026 77680-028 77679-958 77680-002 77680-004 77680-006 77680-010 77680-012 77680-014 77680-018 77680-020
NanoXact Gold Nanospheres, 0.05 mg/ml (Au), Citrate Surface, nanoComposix
Beads Microcarriers
High quality gold nanoparticles. Displaceable citrate surface. Provided with full COA for great value.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Standard colloidal concentration (0.05 mg/mL, 50 ppm, 0.254 mM, near 1 OD [0.8–1.6 depending on size])
  • Citrate surface is stable and displaceable
Spherical gold nanoparticles, 0.05 mg/mL
Each batch is extensively characterized to ensure it meets the highest quality standards, providing reliable and reproducible results. Products are suppliedMore Product Information