NP-40 Alternative, MilliporeSigma
Supplier: EMD MILLIPORE (BIOSCIENCES)
492016-100ML 492016-500ML 492016-1000ML
80601-882EA 108.13 USD
80601-882 80601-884 80603-416
NP-40 Alternative, MilliporeSigma
Protein Extraction Protein Extraction Reagents
NP-40 alternative is a non-ionic surfactant used in the isolation and purification of functional membrane protein complexes and in two-dimensional (2D) gel electrophoresis.