Monoject™ Hypodermic Needles, Sterile, Covidien
Supplier: Cardinal Health
8881250313 1188822112 1188823100 8881200342 8881200326 8881250255 8881200037 1188826112 8881200433 8881250081 8881250206 8881250362 8881200052 1188818112 8881200078 8881250073 8881250289 8881200573 8881200029 8881200466 8881200805 8881250024 8881250040 8881250172 1188819100 1188819112 1188820100 1188825058 1188825100 1188825112 1188830340 8881200318 8881250222 8881250230 8881250107 8881250123 1188818113 1188820112 1188827012 8881200011 8881250016
14222-162CS 422.54 USD
14222-162 82002-366 82002-374 MJ8881-200342 15141-136 15141-190 53498-476 82002-384 MJ8881-200433 MJ8881-250081 MJ8881-250206 15141-192 53498-498 82002-334 MJ8881-200078 MJ8881-250073 MJ8881-250289 14222-306 15141-132 15141-138 15141-140 15141-178 15141-182 15141-188 82002-338 82002-342 82002-346 82002-376 82002-378 82002-380 82002-388 MJ8881-200318 MJ8881-250222 MJ8881-250230 15141-184 15141-186 82002-336 82002-350 82002-386 15141-130 MJ8881-250016
Monoject™ Hypodermic Needles, Sterile, Covidien
Syringe Needles
Ultra-sharp, stainless steel needles are tri beveled, latex free, and anti coring.
  • Needles Available with Aluminum or Polypropylene Luer-Lock Hubs
  • Color Coding
  • Latex Free
Needles are available with aluminum or polypropylene Luer-lock hubs
Rigid pack needles feature a color-coded cap for identification. SoftPack needles feature a one-step peel-away package that facilitates quick and easy opening for aseptic handling and an improved lubricant for increased patient comfort. They also contain a color-coded gauge stripe and bar code on each individualMore Product Information