BLOK™ BSA, 10% BSA Protein Blocking Solution, G-Biosciences
Supplier: G-BIOSCIENCES
786-193 786-195
82022-632EA 281.66 USD
82022-632 76286-966
BLOK™ BSA, 10% BSA Protein Blocking Solution, G-Biosciences
Buffers Electrophoresis Buffers Blocking Buffers
G-Biosciences' BLOK™ BSA is a 10% BSA protein blocking solution suitable for blocking Western blots, ELISA and dot Blots
G-Biosciences' BLOK™ BSA in TBS and in PBS do not contain endogenous biotin; therefore, they are compatible with biotin/avidin systems and result in high signal-to-noise ratios.
Supplied in ready-to-use form, G-Biosciences offers BLOK™ BSA in your choice of TBS or PBS buffers.
Información para pedidos: Accessories available.