KIMBLE® KONTES® Buchner Funnels, with KIMFLOW® Fritted Disc, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
28400-153 28400-303 28400-1503 28400-20002 28400-30001 28400-30002 28400-23 28400-3503 28400-6003 28400-20001 28400-21 28400-22 28400-152 28400-301 28400-302 28400-1502 28400-6002 28400-151 28400-601 28400-602 28400-603 28400-1501 28400-3501 28400-3502 28400-6001
89001-036CS 114.57 USD
89001-036 89001-038 89001-040 89001-042 89001-044 89001-046 89001-372 89001-388 89001-392 89001-394 89000-810 89000-812 89000-814 89000-816 89000-818 89000-820 89000-822 89001-374 89001-376 89001-378 89001-380 89001-382 89001-384 89001-386 89001-390
KIMBLE® KONTES® Buchner Funnels, with KIMFLOW® Fritted Disc, Kimble Chase
Funnels Filtration Funnels Büchner Funnels
For use wherever filter paper and membranes cannot withstand the chemical attack of the filtrate.
  • Has a marking spot
Funnels are constructed with heavy walled stems and beaded top rims for extra strength.
Certificaciones: Designed from Federal Specification DD-F-776. Low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Información de suministro: Supplied with KIMFLOW® fritted disc.