PYREX® VISTA™ Micro Filtering Flasks, Corning
Supplier: Corning
70360-50
89090-314CS 354.94 USD InStock
89090-314 89090-358 89090-396
PYREX® VISTA™ Micro Filtering Flasks, Corning
Flasks Filter Flasks
Made of borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These small filtering flasks are recommended for microchemical use. All flasks have permanent blue enamel marking spots.
Tubulation outer Ø: 10 mm
Certificaciones: ASTM E-1406 Type 2
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty