VWR® Entrance Mats with Stabilizing Frame
Supplier: VWR International
89097-490 89097-492 89097-494 89097-496 89097-498 89097-500 89097-502 89097-504 89097-506 89097-508 89097-510 89097-512
89097-490EA 128.3 USD InStock
89097-490 89097-492 89097-494 89097-496 89097-498 89097-500 89097-502 89097-504 89097-506 89097-508 89097-510 89097-512
VWR® Entrance Mats with Stabilizing Frame
Floor Mats Tacky Mats
Entryway contamination control systems consist of 30- or 60-layered mats applied to a reusable white polystyrene frame.
  • Nonskid backing
  • Effectively captures soil, dirt, and dust
  • Frames are reusable
  • Mats can be easily removed while cleaning
Each layer is coated with an adhesive formulated to capture soil and contain it until the layer is peeled away and discarded
Placing mats in high-traffic areas and entryways will significantly decrease the amount of dust and dirt that typically tracks in from foot traffic and equipment wheels, reduce maintenanceMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty