Luer Valves and Injection Caps, SAI Infusion
Supplier: SAI Infusion Technologies
LLV
89134-222PK 77.11 USD
89134-222
Luer Valves and Injection Caps, SAI Infusion
Valves Injection Valves
SAI offers a variety of Luer Valves and Swabbable Injection Caps to simplify catheter access and prevent the introduction of microbial pathogens.
  • Male Luer for easy catheter or harness connection
  • All parts provided ETO-exposed
These caps can be used anywhere that catheter access is needed, such as harnesses, extension lines, catheters, and Huber needles.
Needle less injection site with male Luer lock are made of polycarbonate, silicone.
Envase: Box of 10.
Order Now
[{"stiboOrderingTable":"<!--Specifications Tab-->\r\n\t\t\t\t<h2>SPECIFICATIONS</h2>\r\n\t\t\t\t <div id="stiboSpecGrid">\r\n\t\t\t\t\t<table id="stiboSpecGridTable" width="100%" class="productGrid">\r\n\t\t\t\t\t\t<caption class="caption">\r\n\t\t\t\t\t\t</caption>\r\n\t\t\t\t\t\t<tbody>\r\n\t\t\t\t\t\t\t<tr>\r\n\t\t\t\t\t\t\t\t<th></th>\t\t\t\t\t\t\t\t\t\t\t\r\n\t\t\t\t\t\t\t</tr>\r\n\t\t\t\t\t\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Environmentally Preferable</td>\r\n\t\t\t<td class="productGrid bld" colspan="1"></td>\r\n\t\t\t</tr>\r\n\t\t<style type="text/css">\r\n \t.stibospecprodgrid {\r\n \t\t\tdisplay:block;\r\n \t\t\toverflow-x:auto;\r\n \t\t\t}\t\r\n</style>\r\n\r\n<script type="text/javascript">\r\n\r\n\t$(document).ready(function() {\r\n\t\tif (numCol("#stiboSpecGridTable") > 4){\r\n\t\t\t$('#stiboSpecGrid').addClass('stibospecprodgrid');\r\n\t\t}\t\t\r\n\t});\r\n\t\r\n\tfunction numCol(table) {\r\n\t var maxColNum = 0;\r\n\t var i=0;\r\n\t var trs = $(table).find("tr");\r\n\t for ( i=0; i<trs.length; i++ ) {\r\n\t maxColNum = Math.max(maxColNum, getColForTr(trs[i]));\r\n\t }\r\n\r\n\t return maxColNum;\r\n\t}\r\n\tfunction getColForTr(tr) {\r\n\r\n\t var tds = $(tr).find("td");\r\n\r\n\t var numCols = 0;\r\n\r\n\t var i=1;\r\n\t for ( i=1; i<tds.length; i++ ) {\r\n\t var span = $(tds[i]).attr("colspan");\r\n\r\n\t if ( span )\r\n\t numCols += parseInt(span);\r\n\t else {\r\n\t numCols++;\r\n\t }\r\n\t }\r\n\t return numCols;\r\n\t}\r\n</script>\t\t\t\t\t\t\t\t\r\n</tbody>\r\n\t\t\t\t\t</table>\r\n\t\t\t\t\t</div>"}]