VWR® Ergonomic Economy Smooth Floor Mats, Black
Supplier: VWR International
89185-440 89185-442 89185-444 89185-446 89185-448 89185-450 89185-452 89185-476 89185-478 89185-480 89185-482 89185-484 89185-486 89185-488 89185-490 89185-510 89185-512 89185-514 89185-516 89185-518 89185-520 89185-524
89185-440EA 120.16 USD InStock
89185-440 89185-442 89185-444 89185-446 89185-448 89185-450 89185-452 89185-476 89185-478 89185-480 89185-482 89185-484 89185-486 89185-488 89185-490 89185-510 89185-512 89185-514 89185-516 89185-518 89185-520 89185-524
VWR® Ergonomic Economy Smooth Floor Mats, Black
Floor Mats Traction and Anti-Fatigue Floor Mats
The VWR® Economy Smooth Mat is a low-density polyurethane material with an anti-fatigue profile. This flat mat is ideal for dry, non-critical environments combining both sitting and standing work areas. No smooth matting available delivers as much relief and comfort at this price. The VWR® Economy Smooth Mat can be customized to any size and any shape, always in one piece.
  • Stimulate the body to increase blood circulation and alertness
  • Molded beveled edges eliminate tripping hazards
  • Silcone free, latex free
  • Easy-to-clean, closed-cell/nonporous top surface
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty