PIG® Absorbent Mat Roll, New Pig
Supplier: New Pig
MAT202-01 MAT220 MAT411 MAT137 MAT245 MAT153 MAT193 MAT225 MAT246 MAT2009
56620-030EA 244.77 USD
56620-030 56620-038 56620-042 56620-196 56620-796 89073-818 89073-830 89073-832 89205-892 89205-894
PIG® Absorbent Mat Roll, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind.
  • Eight layers of 100% polypropylene thermally bonded to be exceptionally strong
  • Won't rip, tear or fray even when saturated
  • Dimple pattern speeds wicking of liquid for faster, easier cleanup
  • Highly absorbent, fine-fiber construction won't leave
More Product Information