PYREX® Fiber Glass Roving, Corning
Supplier: Corning
3950
89219-589CS 2095.93 USD
89219-589
PYREX® Fiber Glass Roving, Corning
Glass and Cotton Wool
This fiber glass is made from fine glass fibers with diameters controlled to approximately 0.008 mm. This makes the glass pliable and easy to use. It contains minimal heavy metals and is suitable for use as a filtering medium.
Manufactured from No. 9989 glass.
Información para pedidos: This product is only sold in a case qty of 2.72 kg (6 lbs.).
Envase: Supplied in the form of roving approximately 2" in diameter by 22' long and packaged in 0.23 kg (½ lbs.) bags.