illustra Ready-To-Go GenomiPhi HY DNA Amplification Kits, Cytiva
Supplier: GLOBAL LIFE SCIENCES SOLUTIONS
25-6603-24 25-6603-96
89233-922EA 686.57 USD
89233-922 89233-924 89233-926
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kits, Cytiva
Nucleic Acid Reagents Isothermal Amplification Reagents
illustra™ Ready-To-Go™ GenomiPhi™ HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample.
  • Representative amplification of the whole genome
  • Preformulated, predispensed, single dose, lyophilised cakes for rapid reaction set-up and greater reproducibility
  • Quick and simple automation-friendly protocol; no thermal cycler required
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint
  • Yields high quality DNA (eg. for high-throughput genotyping and DNA archival)