Foil Rolls
Supplier: Heathrow Scientific
HS23534A HS23536A
89234-756EA 22.09 USD InStock
89234-756 89234-758
Foil Rolls
Aluminum Foil
Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes, and petri dishes.
  • Create custom-size pieces of foil
  • Two sizes available
These foil rolls are ideal for autoclaving, freezing, incubating, protecting lab ware, lining, sealing, and general storage.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty