Base Excision Repair Antibody Panel 2 Kit, Gene Tex
Supplier: Trace Measurement Limited
GTX300021
89329-734EA 836.11 USD
89329-734
Base Excision Repair Antibody Panel 2 Kit, Gene Tex
Assays Nucleic Acid Assays
These sampler kits with combinations of high quality antibodies are convenient and efficient tools to study the mechanism of DNA repair.
Información de suministro: Kit includes 25 µL each of APE1 antibody [N1] (N-term), PARP antibody [N1N2] (N-term), XRCC1 antibody [N1N3], PNK antibody [N1C1], DNA ligase III antibody [C2C3] (C-term) and Rabbit IgG antibody (HRP).
Precaución: Not for any clinical, therapeutic, or diagnostic use in humans or animals. Not for animal or human consumption.