Anti-IgG Bovine Polyclonal Antibody (CF™ 568)

Supplier: Biotium
20294-1MG
89493-408EA 266.7 USD
89493-408
Anti-IgG Bovine Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 568
Epitope:
Host: Bovine
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR