Difco™ Anaerobic Agar, BD Diagnostics
Supplier: Seca Med.Waagen u.Meßsysteme
253610
90003-474EA 506.59 USD
90003-474
Difco™ Anaerobic Agar, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Anaerobic Agar is used for cultivating anaerobic microorganisms.
  • Peptone provides the nitrogen, vitamins, and amino acids
  • Dextrose is a carbon source
  • Sodium chloride maintains the osmotic equilibrium
  • Sodium thioglycollate and sodium formaldehyde sulfoxylate are reducing agents
  • Methylene blue serves as indicator of oxygen production
  • Agar is the solidifying agent
Methylene blue serves as an indicator of anaerobiosis with a blue color indicating the presence of oxygen.