Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Pyruvate dehydrogenase Kinase 1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-PK112-D
95043-892EA 222.36 USD InStock
95043-892
Anti-Pyruvate dehydrogenase Kinase 1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Pyruvate dehydrogenase kinase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Rabbit, Pig, Monkey, Dog

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty