Immobiline™ DryStrip Gels, Cytiva
Supplier: GE Healthcare - Life Sciences
17-1233-01 17-1234-01 17-6001-13 17-6001-14 17-6001-15 17-6002-46 17-6001-10 17-6001-12 17-6001-94 17-6001-96 17-6002-47 17-6003-77 17-6003-74 18-1016-60 18-1016-61 17-1235-01 17-6003-75 17-6003-76 17-6001-97 17-6002-44 17-6002-45 17-6001-11 17-6003-73 17-6001-95
95056-410PK 218.53 USD InStock
95056-410 95056-412 95056-416 95056-418 95056-420 95056-366 95056-380 95056-384 95056-546 95056-552 95056-368 95056-378 95056-404 95056-406 95056-408 95056-414 95056-438 95056-440 95056-554 95056-362 95056-364 95056-382 95056-402 95056-548
Immobiline™ DryStrip Gels, Cytiva
Electrophoresis Gels Precast Gels
Immobiline™ DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application. IPG strips contain a preformed pH gradient immobilised in precast polyacrylamide gels on a plastic backing. A comprehensive range of overlapping IPG strips covering narrow, medium, and wide pH ranges are available in various strip lengths, and the strips are individually packaged and bar coded for convenient handling and sample tracking.
  • Improved Immobiline™ DryStrip gels provide increased stability, reproducibility, and protein
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty