Surgipath® Cassette Storage Drawers, Leica® Microsystems
Supplier: Leica Biosystems
39MC-900-1
95057-864CS 147.65 USD InStock
95057-864
Surgipath® Cassette Storage Drawers, Leica® Microsystems
Boxes Storage Boxes
Drawers offer an efficient means of organizing and filing histology case blocks for future reference.
  • Efficient organization
  • High polymer resin
  • Houses 256 cassettes
Constructed of a high polymer plastic resin, the storage system houses 256 standard cassettes and approximately 500 mini cassettes. Each two drawer system can be used individually for transport, or combined with additional drawers and stacked to create a larger system.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty