Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: KPL
5220-0364
95058-782EA 158.54 USD
95058-782
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Goat Anti-IgG (H+L) Reacts with Rat

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

SPECIFICATIONS

Antigen Symbol IgG
Environmentally Preferable
Antigen Name Immunoglobulin G
Antibody Type Secondary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Rat
Host Goat
Isotype
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow Cytometry Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR