Wash Buffer, Tris Buffered Saline (TBS), 10X, Sera Care
Supplier: MetaSystems Hard&Software GmbH
5460-0037 5460-0036
95059-270EA 516.88 USD
95059-270 95059-268
Wash Buffer, Tris Buffered Saline (TBS), 10X, Sera Care
Buffers Electrophoresis Buffers Washing Buffers
10X Tris-buffered Saline (TBS) is used as a general wash buffer in immunoassay applications.