McIlwain™ Tissue Chopper, Stoelting
Supplier: Stoelting
51350
95060-164EA 4685.26 USD InStock
95060-164
McIlwain™ Tissue Chopper, Stoelting
Microtomes and Microtome Assemblies Microtomes
Tissue Chopper is a proven instrument for rapidly sectioning living or fixed tissue without freezing or embedding to harden the tissue.
  • With chopping Speed of 0-200 strokes/min
  • Can be used in studies of metabolism or for electrophysiology
  • For rapid serial sectioning of multiple preparations of living tissue
Tissue Chopper may also be used to cube fixed tissue for araldite embedding and EM
The table moves at a pre-set speed. As it moves, a chopping arm is raised automatically, and chops down at frequencies from zero to 200 strokes per minute. A pre-More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty