Lab Digital Salinity Refractometer, Sper Scientific
Supplier: Sper Scientific
300035
97011-860EA 932.1 USD
97011-860
Lab Digital Salinity Refractometer, Sper Scientific
Refractometers Benchtop Refractometers
Measure concentrations in brix, salinity, refractive index, and clinical parameters with the full scale accuracy of 0.1% using CCD scanner (linear scanned array imaging) technology
These simple to use models calibrate to distilled water and require a sample of only 0.4mL. Instantly displays temperature in addition to multiple scales with the unit of measure indicated on the large LCD. The sophisticated microprocessor provides automatic calibration, automatic temperature compensation, automatic shutoff (after 3 minutes), and visual min./max. alarms. Low power consumption enables 5,000More Product Information