Sample Loops for Rheodyne Valves, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N9306024 N9306030 N9306029 N9306032 N9306037 N9306033 N9306035 N9306034 N9306041 N9306036 N9306038
97012-974EA 283.43 USD
97012-974 97012-976 97012-978 97012-980 97012-984 97012-986 97012-996 97013-196 97012-972 97012-982 97012-988 97012-990 97012-992 97013-000 97013-194
Sample Loops for Rheodyne Valves, PerkinElmer
Chromatography Supplies Chromatography Sample Loops
Sample loops are often used to absorb sample components resulting in inaccurate quantitative results. In addition, metal loops may be corroded by high-salt buffers and chloride salts. Sample Loops are made from PEEK tubing and fittings.
PEEK (Poly-Ether-Ether Ketone) is a mechanically strong, chemically inert polymer ideal for HPLC applications where metal surfaces may interact with mobile phase or sample components.
Loops are compatible with both manual valves and Series 200 Autosampler valves.
Información para pedidos: Each Sample Loop is supplied with two PEEK hex-head nuts andMore Product Information