Zero Dilution Inner Liner, 2 ext.Ø.×73L mm, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N1011446
97019-184EA 165.16 USD InStock
97019-184
Zero Dilution Inner Liner, 2 ext.Ø.×73L mm, PerkinElmer
Chromatography Supplies Chromatography Inlet Liners
To be used in conjunction with outer liner (N1011447).
  • Not compatible with the Clarus 590/690 Capillary Injectors
  • 73 mm length
Inlet liners for split injection have mixing chambers with tortuous flow paths to allow full vaporization of the sample. Deactivating the surface of these liners prevents active compounds from degrading. Packing the liner with wool will trap non-volatile residue and prevent column contamination when analyzing dirty samples.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty