Benchtop Conductivity/TDS/Salinity Meter, Sper Scientific
Supplier: Sper Scientific
860032
97039-650EA 457.62 USD
97039-650
Benchtop Conductivity/TDS/Salinity Meter, Sper Scientific
Electrochemistry Meters Conductivity Meters
Rugged housing with large LCD displaying the parameter being measured together with time, date and temperature (in °C or °F).
  • Automatic or manual temperature compensation
  • Automatic or manual ranging, automatic buffer recognition
  • Digital and analog outputs
  • Min/max and 99 data points
  • Data hold
  • 5 calibration points
  • Electrode status indication
Operates on AC voltage (no batteries).
Información para pedidos: 5-year warranty. Comes ready to use with ATC conductivity electrode, electrode holder arm, AC adaptor, computer cable and software.