VWR® Phosphate Buffer Solution
Supplier: VWR Solon Production
E715-1L
97062-958EA 136.36 USD
97062-958 97062-960
VWR® Phosphate Buffer Solution
Buffers Cell Culture Buffers
Phosphate Buffered Saline (PBS) with 0.05% Tween® 20, pH 7.5 is a ready-to-use 1X concentration