Thiourea ≥99%, Proteomics Grade

Supplier: VWR Solon Production
Warning

M226-100G
97064-928CS 1156.35 USD
97064-928 97064-930
Thiourea ≥99%, Proteomics Grade
Thiourea
Formula: NH₂CSNH₂
MW: 76.12 g/mol
Melting Pt: 171 °C…184 °C
Density: 1.435 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9 III
Merck Index: 13,09443
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Melting Range 165°C - 178°C
Protease none detected
Purity > =99 %
Solubility (1%, Water) Pass
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR