Avantor® Seradigm, Premium Grade Fetal Bovine Serum (FBS)
Supplier: VWR
1500-500G 1500-500GH 1500-050 1500-050H 1500-500 1500-500H 1500-050T 1500-050TH 1500-500T 1500-500TG 1500-500TH
97068-086EA 798.84 USD
97068-086 97068-088 89510-194 89510-196 97068-085 97068-091 76308-942 76308-944 76308-946 76308-982 76308-984
Avantor® Seradigm, Premium Grade Fetal Bovine Serum (FBS)
Sera (Serum)
Premium Grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a wide variety of applications.
  • 100% US Origin
  • High quality, high performance FBS for a variety of applications
  • Preservation of native, growth-promoting factors and minimization of growth-inhibiting factors
  • ISIA Certified Supply Chain for Traceability
The care and attention we devote to the manufacture of our products generate final test results that are substantially lower than product specifications
Avantor Seradigm is a supplier of animal seraMore Product Information