Praseodymium 99% REO basis, foil, Thickness: 0.25 mm (0.01 in)

Supplier: Origen Biomedical, Inc

010305-FF
AA10305-FFEA 493.5 USD
AA10305-FF
Praseodymium 99% REO basis, foil, Thickness: 0.25 mm (0.01 in)
Praseodymium

MDL: MFCD00011174


Purity: 99.5% CAS No.: 7440-10-0 Physical Form: ≈1.27g/25x25mm, Packaged in mylar under argon Appearance: Gray Odor: Odorless MDL No.: MFCD00011174

Formula: Pr
MW: 140.91
Storage Temperature: Ambient
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR