Diethyl mesoxalate (diethyl ketomalonate) 95%

Supplier: Origen Biomedical, Inc
Warning

A12771-22 A12771-14 A12771-06
AAA12771-22EA 496 USD
AAA12771-22 AAA12771-14 AAA12771-06
Diethyl mesoxalate (diethyl ketomalonate) 95%
Diethyl mesoxalate (diethyl ketomalonate)

Fieser: 6,188 8,170 10,143


Purity: 95% CAS No.: 609-09-6 Molecular Formula: C7H10O5 Formula Weight: 174.15 Flash Point: 110°(230°F) Refractive Index: 1.4150 Melting Point: ca -70° Boiling Point: 208-210° Beilstein Reference: 971873 Merck Reference: 14,5913 MDL No.: MFCD00009121

Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 208…210 °C
Melting Pt: ∼ …70 °C
Density: 1.142
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009121
CAS Number: 609-09-6
EINECS: 210-176-2
Merck Index: 14,5913

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR