Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fumaronitrile 98+%

Supplier: Origen Biomedical, Inc
Danger

L14911-06
AAAL14911-06EA 76.8 USD InStock
AAAL14911-06
Fumaronitrile 98+%
Fumaronitrile
Purity: 98+% CAS No.: 764-42-1 Molecular Formula: C4H2N2 Formula Weight: 78.07 Odor: Recognizable Melting Point: 94-99° Boiling Point: 186° Beilstein Reference: 969245 UN No.: 3439 MDL No.: MFCD00001928


Formula: C₄H₂N₂
MW: 78.07 g/mol
Boiling Pt: 186
Melting Pt: 96
Density: 0.94
Storage Temperature: Ambient
MDL Number: MFCD00001928
CAS Number: 764-42-1
EINECS: 212-122-3
UN: 3439
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty