Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoro-1,3,5-trimethylcyclohexane mixture of isomers, tech.

Supplier: Origen Biomedical, Inc

B21500-18
AAB21500-09EA 50.36 USD InStock
AAB21500-09 AAB21500-18
Perfluoro-1,3,5-trimethylcyclohexane mixture of isomers, tech.
Perfluoro-1,3,5-trimethylcyclohexane
CAS No.: 374-76-5 Molecular Formula: C9F18 Formula Weight: 450.07 Refractive Index: 1.2970 Melting Point: -68° Boiling Point: 125-128° MDL No.: MFCD00066614


Formula: C₉F₁₈
MW: 450.07 g/mol
Boiling Pt: 125…128 °C
Melting Pt: –68 °C
Density: 1.888
Storage Temperature: Ambient
MDL Number: MFCD00066614
CAS Number: 374-76-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty