1,6-Bis(diphenylphosphino)hexane 97%

Supplier: Origen Biomedical, Inc
Warning

H55528-03 H55528-06 H55528-14
AAH55528-03EA 30.3 USD
AAH55528-03 AAH55528-06 AAH55528-14
1,6-Bis(diphenylphosphino)hexane 97%
1,6-Bis(diphenylphosphino)hexane

Purity: 97% CAS No.: 19845-69-3 Molecular Formula: C30H32P2 Formula Weight: 454.53 Melting Point: 124-126° MDL No.: MFCD00003053

Formula: C₃₀H₃₂P₂
MW: 454.53 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00003053
CAS Number: 19845-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR